Learn More
Enzo Life Sciences HSP 90 (human), (native) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPP770D
Encompass_Preferred
Alternative name: HSP86, Heat shock protein 90. MW: ~90kDa. Purity: ≥90% (SDS-PAGE; Western blot). Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 1.0mM DTT and 100mM sodium chloride. Source: Produced in HeLa cells. UniProt: P07900 (HSP90α), P08238 (HSP90β). Application: ELISA, WB, Antigen microarray. Recommended dilutions: Western Blot (100ng, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Long Term Storage: -80°C
Catalog No. 50-667-772