Learn More
Enzo Life Sciences HSP90 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA836F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 250-325 of human Hsp90. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Rabbit. Applications: IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7542