Learn More
Enzo Life Sciences HSP90alpha polyclonal antibody (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPS771F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of mouse Hsp86. UniProt: P07901. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Fish, Dog, Bovine, Beluga. Applications: IHC, WB, Inhibition assay, Cell migration. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide and 50% glycerol. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-790