Learn More
Enzo Life Sciences HSP 90 polyclonal antibody (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA846F
Encompass_Preferred
Host: Rabbit. Immunogen: Human Hsp90. UniProt: P07900 (HSP90alpha), P08238 (HSP90beta). Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Bovine. Applications: IHC (PS), WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-641