Learn More
Enzo Life Sciences HSP90alpha monoclonal antibody (9D2) (HRP conjugate) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA840HRPF
Encompass_Preferred
Clone: 9D2. Host: Rat. Isotype: IgG2a. Immunogen: Purified Hsp90 isolated from human therapeutic orchiectomy specimens. Source: Purified from mouse ascites. Species reactivity: Human Chicken. Applications: WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide diluted in proprietary stabilizer solution.
Catalog No. 50-200-7550