Learn More
Enzo Life Sciences HSP90alpha monoclonal antibody (9D2) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA840D
Encompass_Preferred
Clone: 9D2. Host: Rat. Isotype: IgG2a. Immunogen: Purified Hsp90 isolated from human therapeutic orchiectomy specimens. UniProt ID: P07900. Source: Purified from mouse ascites. Species reactivity: Human Chicken. Applications: Flow Cytometry, ICC, IHC, IP, WB. Recommended Dilutions: Flow Cytometry (1:100) Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7547