Learn More
Enzo Life Sciences HSP90alpha monoclonal antibody (K41009) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA839050
Encompass_Preferred
Clone: K41009. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant human Hsp90α. UniProt ID: P07900. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat, Rabbit. Applications: WB. Recommended Dilutions: Western Blot (1µg/ml, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7543