Learn More
Enzo Life Sciences HSP90alpha polyclonal antibody (PE conjugate) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPS771PE050
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of mouse Hsp86. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Fish, Dog, Bovine, Beluga. Applications: Flow Cytometry. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid vigorous centrifugation and vortexing. Do not freeze. Protect from light.
Catalog No. 50-200-7658