Learn More
BPS Bioscience Inc HSP90β (C-terminal), Biotin-Labeled, His-Tag, Avi-Tag Recombinant, 100 µg
SDP
Catalog No. NC2737911
Encompass
Recombinant human HSP90β (Human Heat Shock Protein 90 beta), encompassing amino acids 527-724(end). This construct contains an N-terminal His-tag (6xHis) followed by an Avi-tag™. The recombinant protein was enzymatically biotinylated at the C-terminal using Avi-Tag technology and affinity purified. Biotinylation is confirmed to be ≥90%.Synonyms - Hsp90beta, heat shock protein 90 beta
Catalog No. NC2737911 Supplier BPS Bioscience Inc Supplier No. 50313
Recombinant human HSP90β (Human Heat Shock Protein 90 beta), encompassing amino acids 527-724(end). This construct contains an N-terminal His-tag (6xHis) followed by an Avi-tag™. The recombinant protein was enzymatically biotinylated at the C-terminal using Avi-Tag technology and affinity purified. Biotinylation is confirmed to be ≥90%.Synonyms - Hsp90beta, heat shock protein 90 beta