Learn More
Aviva Systems Biology Corp HSP90B1 Antibody - C-terminal region (ARP40463_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40463_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSP90B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HSP90B1. Alternate names: ECGP, GP96, GRP94, TRA1ApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57465