Learn More
Enzo Life Sciences HSP90beta (human), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPP777F
Encompass_Preferred
Alternative name: HSP84, Heat shock protein 90beta. Purity: ≥85% (SDS-PAGE; Western blot). MW: ~90kDa. Formulation: Liquid. In 20mM TRIS, pH 7.5, containing 500mM sodium chloride, 5% glycerol, and 1mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7646