Learn More
Enzo Life Sciences HSP90beta monoclonal antibody (K3701) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA843F
Encompass_Preferred
Clone: K3701. Host: Mouse. Isotype: IgM. Immunogen: Recombinant human HSP90β. UniProt ID: P08238. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Hamster, Guinea pig, Dog, Bovine. Applications: Flow Cytometry, WB. Recommended Dilutions: Flow Cytometry (10µg/ml) Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Ammonium sulfate precipitation purified. Formulation: Liquid. In saline, pH 6.9, containing 0.1% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7556