Learn More
Enzo Life Sciences HSP90beta polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA844200
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human HSP90β. Source: Purified from rabbit serum. Species reactivity: Human, Mouse Sheep, Rabbit, Porcine, Hamster, Guinea pig, Dog, Bovine. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7558