Learn More
Enzo Life Sciences HSP90beta polyclonal antibody (PE conjugate) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA844PE200
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human HSP90β. UniProt ID: P08238. Source: Purified from rabbit serum. Species reactivity: Human, Mouse Sheep, Rabbit, Porcine, Hamster, Guinea pig, Dog, Bovine. Applications: Flow Cytometry. Recommended Dilutions: Flow Cytometry (1:100) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles. Protect from light.
Catalog No. 50-200-7560