Learn More
Selleck Chemical LLC HSP990
SDP
Catalog No. 5000002235
Encompass_Preferred
HSP990 (NVP-HSP990) is a novel potent and selective HSP90 inhibitor for HSP90 / with IC50 of 0 6 nM/0 8 nM NVP-HSP990 induces cell cycle arrest and apoptosis
Catalog No. 50-000-02235 Supplier Selleck Chemical LLC Supplier No. S709725MG
HSP990 (NVP-HSP990) is a novel potent and selective HSP90 inhibitor for HSP90 / with IC50 of 0 6 nM/0 8 nM NVP-HSP990 induces cell cycle arrest and apoptosis