Learn More
Selleck Chemical LLC HSP990 (NVP-HSP990) 100mg 934343-74-5
SDP
Supplier:  Selleck Chemical LLC S7097100MG
Encompass_Preferred
HSP990 (NVP-HSP990) is a novel, potent and selective HSP90 inhibitor for HSP90?/? with IC50 of 0.6 nM/0.8 nM. NVP-HSP990 induces cell cycle arrest and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4399