Learn More
Aviva Systems Biology Corp Hspa1a Antibody - N-terminal region (ARP61005_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61005_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hspa1a. It was validated on Western Blot. . Alternate names: Hsp70-3, Hsp70.3, Hsp72, MGC189852, hsp68, hsp70A1ApplicationWB. Species reactivity: Dog, Goat, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-287-229