Learn More
Sino Biological HSPA1A Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11660T54100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HSPA1A (rh HSPA1A; Catalog#11660-H07B; NP_005337.2; Ala2-Asp641). HSPA1A specific IgG was purified by Human HSPA1A affinity chromatography.
Catalog No. 50-224-6372