Learn More
Aviva Systems Biology Corp HSPA1L Antibody - C-terminal region (ARP53636_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53636_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPA1L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HSPA1L. Alternate names: HSP70-HOM, hum70t, HSP70T, HSP70-1LApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-61076