Learn More
Aviva Systems Biology Corp HSPA2 Antibody - middle region (ARP55382_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55382_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HSPA2. Alternate names: HSP70-2, HSP70-3ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61487