Learn More
Aviva Systems Biology Corp HSPA4 Antibody - middle region (ARP46074_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46074_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPA4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HSPA4. Alternate names: APG-2, HS24/P52, MGC131852, RY, hsp70, hsp70RY, HSPH2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59210