Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human HSPA4 (HSP70) ELISA Kit

Catalog No. EHHSPA4
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHHSPA4 96 Tests
EHHSPA4X10 10 x 96 Tests
EHHSPA4X5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EHHSPA4 Supplier Invitrogen™ Supplier No. EHHSPA4
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human HSPA4 (HSP70) ELISA quantitates Hu HSPA4 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu HSPA4. Principle of the method The Human HSPA4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The HSP 70 family is composed of four highly conserved proteins: HSP 70, HSC 70, GRP 75 and GRP 78. HSP 70 expression is strongly induced in response to heat stress. HSP 70 and HSC 70 play key roles in the cytosolic endoplasmic reticulum and mitochondrial import machinery and are found in both the cytosol and nucleus of mammalian cells. GRP 78 is localized in the endoplasmic reticulum, where it receives imported secretory proteins and is involved in the folding and translocation of nascent peptide chains. GRP 75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. It has been postulated that members of the HSP 70 family act as force-generating motors, relying on the hydrolysis of ATP for their activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P34932
Assay Range 2.469-600 ng/mL
Assay Sensitivity 2 ng/mL
Calibration Range 2 ng/mL to 600 ng/mL
Content And Storage Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Mitochondrial Proteins
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody also can recognize mouse and rat HSP70
Sufficient For 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 70kDa; AI317151; APG2; APG-2; epididymis secretory sperm binding protein Li 5a; heat shock 70 kDa protein 4; heat shock 70kD protein 4; heat shock 70kDa protein 4; Heat shock 70-related protein APG-2; Heat shock protein; heat shock protein 4; heat shock protein Apg-2; heat shock protein family A (Hsp70) member 4; heat shock protein, 110 kDa; HEL-S-5a; HS24/P52; Hsp110; hsp70; hsp70 RY; Hsp70RY; Hspa4; HSPH2; Irp94; ischemia responsive 94 kDa protein; mKIAA4025; RY70kDa; AI317151; APG2; APG-2; epididymis secretory sperm binding protein Li 5a; heat shock 70 kDa protein 4; heat shock 70kD protein 4; heat shock 70kDa protein 4; Heat shock 70-related protein APG-2; Heat shock protein; heat shock protein 4; heat shock protein Apg-2; heat shock protein family A (Hsp70) member 4; heat shock protein, 110 kDa; HEL-S-5a; HS24/P52; Hsp110; hsp70; hsp70 RY; Hsp70RY; Hspa4; HSPH2; Irp94; ischemia responsive 94 kDa protein; mKIAA4025; RY; APG-2; HEL-S-5a; HS24/P52; HSPH2; RY; hsp70; hsp70RY
Gene ID (Entrez) 3308
Gene Symbol HSPA4
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases HSP70RY, Heat shock 70-related protein APG-2
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Cell Cycle & Proliferation
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym APG-2, HEL-S-5 A, HS24/P52, HSPH2, RY, hsp70, hsp70RY
Target HSPA4
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.