Learn More
Aviva Systems Biology Corp HSPA4L Antibody - C-terminal region (ARP53693_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53693_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPA4L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HSPA4L. Alternate names: APG-1, Osp94, HSPH3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53725