Learn More
Aviva Systems Biology Corp HSPA8 Antibody - N-terminal region (ARP48446_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48446_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPA8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HSPA8. Alternate names: HSC54, HSC70, HSC71, HSP71, HSP73, HSPA10, LAP1, MGC131511, MGC29929, NIP71ApplicationIHC, WB. Species reactivity: Goat, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-014-59827