Learn More
Aviva Systems Biology Corp HSPA9 Antibody - C-terminal region (ARP30175_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30175_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPA9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HSPA9. Alternate names: CSA, GRP75, HSPA9B, MGC4500, MOT, MOT2, MTHSP75, PBP74, mot-2, GRP-75ApplicationIHC, IP, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54138