Learn More
Aviva Systems Biology Corp HSPB6 Antibody - middle region (ARP48436_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48436_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPB6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HSPB6. Alternate names: FLJ32389, Hsp20ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-53553