Learn More
Aviva Systems Biology Corp Hspb7 Antibody - C-terminal region (ARP41601_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41601_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hspb7. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 27kDa, Hsp25-2, MGC107591, cvHspApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-946