Learn More
Aviva Systems Biology Corp HSPB8 Antibody - N-terminal region (ARP55035_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55035_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPB8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HSPB8. Alternate names: CMT2L, DHMN2, E2IG1, H11, HMN2, HMN2A, HSP22ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-61407