Learn More
ABclonal Technology HSPB8/HSP22 Rabbit mAb
SDP
Catalog No. 5000011758
Encompass_Preferred
The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule The expression of this gene in induced by estrogen in estrogen receptor-positive breast cancer cells and this protein also functions as a chaperone in association with Bag3 a stimulator of macroautophagy Thus this gene appears to be involved in regulation of cell proliferation apoptosis and carcinogenesis and mutations in this gene have been associated with different neuromuscular diseases including Charcot-Marie-Tooth disease
Catalog No. 50-000-11758 Supplier ABclonal Technology Supplier No. A2539620UL
The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule The expression of this gene in induced by estrogen in estrogen receptor-positive breast cancer cells and this protein also functions as a chaperone in association with Bag3 a stimulator of macroautophagy Thus this gene appears to be involved in regulation of cell proliferation apoptosis and carcinogenesis and mutations in this gene have been associated with different neuromuscular diseases including Charcot-Marie-Tooth disease