Learn More
Aviva Systems Biology Corp HSPBAP1 Antibody - middle region (ARP33578_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33578_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPBAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HSPBAP1. Alternate names: FLJ22623, FLJ39386, PASS1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55078