Learn More
Aviva Systems Biology Corp HSPD1 Antibody - C-terminal region (AVARP09014_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09014_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human HSPD1. Alternate names: CPN60, GROEL, HSP60, HSP65, HuCHA60, SPG13, HLD4, HSP-60ApplicationWB. Species reactivity: Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-014-62567