Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3339RTM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: A7L6. Clonality: Monoclonal. Host: Rat. Isotype: IgG2a kappa. Positive Control: Breast carcinomas, squamous cell carcinomas. Cellular Staining: Basement membrane. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Murine EHS laminin preparation. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: >400kDa.Chromosome: 1p36.1-p34. Gene Symbol: HSPG2. Gene ID: 3339. Ensemble: ENSG00000142798. Uniprot: XP_016856610.
Catalog No. 50-188-4787