Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters HSPgel Column, 5 μm, 6 mm × 150 mm, 100 to 1000 MW

Supplier:  Waters Corp 186001773

Encompass_Preferred

These columns are designed for high-speed GPC (Gel Permeation Chromatography) analysis, providing fast, accurate, and precise molecular weight polymer determinations. They increase sample throughput and reduce solvent consumption and disposal costs. These columns enable total analysis times of 7 minutes or less, leading to up to 6x increased sample throughput and a decrease in solvent usage from 35 to 50 mL or less per run.

  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • Molecular weight range: 100 to 1000
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Endfitting type: Parker-style
  • Packing solvent: ODCB
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 6 mm
  • Length: 150 mm
  • Units per package: 1 pk

Catalog No. 50-780-744


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.