Learn More
Waters Corp HSPgel Column, 5 μm Particle Size, 6 mm X 150 mm, 100–1K Molecular Weight
Catalog No. 50780732 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
HSPgel columns are designed for high-speed GPC analysis, providing fast, accurate, and precise molecular weight polymer determinations. They increase sample throughput and reduce solvent consumption, with total analysis times of 7 minutes or less due to lower volume columns and high-performance solvent management systems.
  • Designed for high-speed GPC analysis.
  • Offers fast, accurate, and precise molecular weight polymer determinations.
  • Increases sample throughput and reduces solvent consumption.
  • Achieves total analysis times of 7 minutes or less.
  • Utilizes lower volume columns for efficient analysis.
  • Decreases solvent usage from 35–50 mL to 5 mL or less per run.
  • Minimizes system band broadening.
  • Certificate of analysis and performance test chromatogram included.
  • Not advised to use a guard column; a 0.5 μm in-line filter can be used instead.
Catalog No. 50-780-732 Supplier Waters Corp Supplier No. 186001761
HSPgel columns are designed for high-speed GPC analysis, providing fast, accurate, and precise molecular weight polymer determinations. They increase sample throughput and reduce solvent consumption, with total analysis times of 7 minutes or less due to lower volume columns and high-performance solvent management systems.
  • Designed for high-speed GPC analysis.
  • Offers fast, accurate, and precise molecular weight polymer determinations.
  • Increases sample throughput and reduces solvent consumption.
  • Achieves total analysis times of 7 minutes or less.
  • Utilizes lower volume columns for efficient analysis.
  • Decreases solvent usage from 35–50 mL to 5 mL or less per run.
  • Minimizes system band broadening.
  • Certificate of analysis and performance test chromatogram included.
  • Not advised to use a guard column; a 0.5 μm in-line filter can be used instead.