Learn More
Waters Corp Waters HSPgel HT 2.0 Column, 5 μm, 500 to 10,000 MW
Supplier:  Waters Corp 186001762
Encompass_Preferred
This HSPgel HT 2.0 column is designed for organic Gel Permeation Chromatography (GPC) conducted between room temperature and 180°C. It is shipped packed in Tetrahydrofuran (THF) and can withstand multiple solvent switches.
  • Designed for organic GPC conducted between room temperature and 180°C.
  • Shipped packed in Tetrahydrofuran (THF).
  • Withstands multiple solvent switches.
Catalog No. 50-780-733