Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters HSPgel Column HT 2.5, 5 μm, 6 mm X 150 mm, ODCB

Supplier:  Waters Corp 186001775

Encompass_Preferred

The new HSPgel columns are engineered for high-speed Gel Permeation Chromatography (GPC) analysis. They are designed to deliver fast, accurate, and precise molecular weight polymer determinations, significantly increasing sample throughput and reducing solvent consumption.

  • Engineered for high-speed Gel Permeation Chromatography (GPC) analysis
  • Delivers fast, accurate, and precise molecular weight polymer determinations
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • Molecular weight range: 1,000 to 20,000
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Endfitting type: Parker-style
  • Packing solvent: ODCB
  • Compatible with HPLC systems and GPC technique
  • USP classification: L21
  • Dimensions: 6 mm inner diameter, 150 mm length
  • Supplied as 1 pk

Catalog No. 50-780-746


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.