Learn More
Waters Corp HPLC Column, HSPgel, 5 μm, 6 x 150 mm, THF
Catalog No. 50780734 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The new HSPgel columns are specifically designed for high-speed GPC analysis. They provide fast, accurate, and precise molecular weight polymer determinations. Additionally, they offer increased sample throughput and significantly reduce solvent consumption and disposal costs. These columns allow for total analysis times of 7 minutes or less, leading to up to a 6× increase in sample throughput and a reduction in solvent usage from 35-50 mL to 5 mL or less per run.
  • High-speed GPC analysis
  • Fast, accurate, and precise molecular weight polymer determinations
  • Increased sample throughput (up to 6×)
  • Greatly reduced solvent consumption (to 5 mL or less per run)
  • Reduced solvent disposal costs
  • Total analysis times of 7 minutes or less
Catalog No. 50-780-734 Supplier Waters Corp Supplier No. 186001763
The new HSPgel columns are specifically designed for high-speed GPC analysis. They provide fast, accurate, and precise molecular weight polymer determinations. Additionally, they offer increased sample throughput and significantly reduce solvent consumption and disposal costs. These columns allow for total analysis times of 7 minutes or less, leading to up to a 6× increase in sample throughput and a reduction in solvent usage from 35-50 mL to 5 mL or less per run.
  • High-speed GPC analysis
  • Fast, accurate, and precise molecular weight polymer determinations
  • Increased sample throughput (up to 6×)
  • Greatly reduced solvent consumption (to 5 mL or less per run)
  • Reduced solvent disposal costs
  • Total analysis times of 7 minutes or less