Learn More
Waters Corp HSPgel Column, ODCB, 5 μm, 6 mm x 150 mm
Catalog No. 50780747 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
HSPgel columns are designed for high-speed GPC analysis, offering fast, accurate, and precise molecular weight polymer determinations. They enable increased sample throughput, reduced solvent consumption, and lower disposal costs. These lower-volume columns provide accurate, reproducible analysis in 7 minutes or less.
  • Enables fast, accurate, and precise molecular weight polymer determinations.
  • Boosts sample throughput by up to 6x.
  • Reduces solvent consumption from 35 to 50 mL to 5 mL or less per run.
  • Lowers solvent disposal costs.
  • Ensures accurate and reproducible polymer analysis with lower volume columns.
  • Completes analysis in 7 minutes or less.
Catalog No. 50-780-747 Supplier Waters Corp Supplier No. 186001776
HSPgel columns are designed for high-speed GPC analysis, offering fast, accurate, and precise molecular weight polymer determinations. They enable increased sample throughput, reduced solvent consumption, and lower disposal costs. These lower-volume columns provide accurate, reproducible analysis in 7 minutes or less.
  • Enables fast, accurate, and precise molecular weight polymer determinations.
  • Boosts sample throughput by up to 6x.
  • Reduces solvent consumption from 35 to 50 mL to 5 mL or less per run.
  • Lowers solvent disposal costs.
  • Ensures accurate and reproducible polymer analysis with lower volume columns.
  • Completes analysis in 7 minutes or less.