Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Column, 5 μm, 6 mm X 150 mm, 10K to 400K

Catalog No. 50780748 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These columns are designed for high-speed GPC analysis, offering fast, accurate, and precise molecular weight polymer determinations. They increase sample throughput and reduce solvent consumption, with analysis times under 7 minutes and solvent usage as low as 5 mL per run.

  • High-speed GPC analysis
  • Fast, accurate, precise molecular weight polymer determinations
  • Increased sample throughput; reduced solvent consumption
  • Analysis times under 7 minutes
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • Molecular weight range: 10,000 to 400,000
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Endfitting type: Parker-style
  • Packing solvent: ODCB
  • System: HPLC
  • Dimensions: 6 mm ID, 150 mm length
  • Units per package: 1 pack

Catalog No. 50-780-748 Supplier Waters Corp Supplier No. 186001777
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These columns are designed for high-speed GPC analysis, offering fast, accurate, and precise molecular weight polymer determinations. They increase sample throughput and reduce solvent consumption, with analysis times under 7 minutes and solvent usage as low as 5 mL per run.

  • High-speed GPC analysis
  • Fast, accurate, precise molecular weight polymer determinations
  • Increased sample throughput; reduced solvent consumption
  • Analysis times under 7 minutes
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • Molecular weight range: 10,000 to 400,000
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Endfitting type: Parker-style
  • Packing solvent: ODCB
  • System: HPLC
  • Dimensions: 6 mm ID, 150 mm length
  • Units per package: 1 pack

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.