Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp HSPgel Column, HT 7, 5 μm, 6.0 x 150 mm

Catalog No. 50780751 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Waters HSPgel Column is an SEC/GPC column designed for organic size exclusion chromatography (GPC) applications. It features a spherical, fully porous polymer matrix and is suitable for high-performance liquid chromatography (HPLC).

  • SEC / GPC columns
  • Length: 150 mm
  • Internal diameter: 6.0 mm
  • Particle size: 5 μm
  • Chemistry: HT 7
  • USP Class: L21
  • Country of origin: United States of America
  • Lower molecular weight: 100,000 (PS)
  • Upper molecular weight: 15,000,000 (PS)
  • Mode of separation: Organic size exclusion chromatography (GPC)
  • Technology: HPLC
  • Matrix: Polymer
  • Particle morphology: Spherical, fully porous

Catalog No. 50-780-751 Supplier Waters Corp Supplier No. 186001780
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Waters HSPgel Column is an SEC/GPC column designed for organic size exclusion chromatography (GPC) applications. It features a spherical, fully porous polymer matrix and is suitable for high-performance liquid chromatography (HPLC).

  • SEC / GPC columns
  • Length: 150 mm
  • Internal diameter: 6.0 mm
  • Particle size: 5 μm
  • Chemistry: HT 7
  • USP Class: L21
  • Country of origin: United States of America
  • Lower molecular weight: 100,000 (PS)
  • Upper molecular weight: 15,000,000 (PS)
  • Mode of separation: Organic size exclusion chromatography (GPC)
  • Technology: HPLC
  • Matrix: Polymer
  • Particle morphology: Spherical, fully porous

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.