Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters HSPgel Column, 5 μm, 6 mm × 150 mm, 100K to 15M

Catalog No. 50780739 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

HSPgel columns are designed for high-speed GPC analysis, providing fast, accurate, and precise molecular weight polymer determinations. They significantly increase sample throughput and reduce solvent consumption and disposal costs. These columns enable accurate and reproducible analysis of polymers using lower volume columns, leading to total analysis times of 7 min. or less, increasing throughput up to 6×, and decreasing solvent usage from 35 to 50 mL to 5 mL or less per run.

  • Packing solvent: THF.
  • System: HPLC.
  • Technique: GPC.
  • USP classification: L21.
  • Particle shape: Spherical.
  • Particle size: 5 μm.
  • Inner diameter: 6 mm.
  • Length: 150 mm.
  • Molecular weight range: 100,000 to 15,000,000.

Catalog No. 50-780-739 Supplier Waters Corp Supplier No. 186001768
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HSPgel columns are designed for high-speed GPC analysis, providing fast, accurate, and precise molecular weight polymer determinations. They significantly increase sample throughput and reduce solvent consumption and disposal costs. These columns enable accurate and reproducible analysis of polymers using lower volume columns, leading to total analysis times of 7 min. or less, increasing throughput up to 6×, and decreasing solvent usage from 35 to 50 mL to 5 mL or less per run.

  • Packing solvent: THF.
  • System: HPLC.
  • Technique: GPC.
  • USP classification: L21.
  • Particle shape: Spherical.
  • Particle size: 5 μm.
  • Inner diameter: 6 mm.
  • Length: 150 mm.
  • Molecular weight range: 100,000 to 15,000,000.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.