Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp HSPgel Column, RT 4.0, 3 μm, 6 mm x 150 mm

Catalog No. 50780725 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This column is designed for high-performance liquid chromatography (HPLC), specifically for size exclusion chromatography (SEC) and organic size exclusion chromatography (GPC). It features a 3 μm spherical, fully porous polymer matrix and is suitable for separating molecules with molecular weights ranging from 10,000 to 400,000 (PS).

  • Product type: SEC / GPC columns
  • Column dimensions: 6.0 mm internal diameter, 150 mm length
  • Particle size: 3 μm
  • Chemistry term: RT 4.0
  • Molecular weight range: 10,000 to 400,000 (PS)
  • Mode of separation: SEC / Organic Size Exclusion Chromatography (GPC)
  • Technology: HPLC
  • Matrix: Polymer, spherical, fully porous

Catalog No. 50-780-725 Supplier Waters Corp Supplier No. 186001753
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This column is designed for high-performance liquid chromatography (HPLC), specifically for size exclusion chromatography (SEC) and organic size exclusion chromatography (GPC). It features a 3 μm spherical, fully porous polymer matrix and is suitable for separating molecules with molecular weights ranging from 10,000 to 400,000 (PS).

  • Product type: SEC / GPC columns
  • Column dimensions: 6.0 mm internal diameter, 150 mm length
  • Particle size: 3 μm
  • Chemistry term: RT 4.0
  • Molecular weight range: 10,000 to 400,000 (PS)
  • Mode of separation: SEC / Organic Size Exclusion Chromatography (GPC)
  • Technology: HPLC
  • Matrix: Polymer, spherical, fully porous

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.