Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters HSPgel Column, 5 μm, 6 mm × 150 mm, 5K – 10M, THF

Catalog No. 50780731 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The HSPgel columns are designed for high-speed GPC analysis, providing fast, accurate, and precise molecular weight polymer determinations. They also offer increased sample throughput and reduced solvent consumption and disposal costs. With modern solvent management systems, many polymers can be analyzed accurately and reproducibly using lower volume columns.

  • Separation mode: SEC/GPC.
  • Molecular weight range: 5000 to 10,000,000.
  • Particle size: 5 μm.
  • Particle shape: Spherical.
  • Endfitting type: Parker-style.
  • Inner diameter: 6 mm.
  • Length: 150 mm.
  • Packing solvent: THF.
  • System: HPLC.

Catalog No. 50-780-731 Supplier Waters Corp Supplier No. 186001760
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The HSPgel columns are designed for high-speed GPC analysis, providing fast, accurate, and precise molecular weight polymer determinations. They also offer increased sample throughput and reduced solvent consumption and disposal costs. With modern solvent management systems, many polymers can be analyzed accurately and reproducibly using lower volume columns.

  • Separation mode: SEC/GPC.
  • Molecular weight range: 5000 to 10,000,000.
  • Particle size: 5 μm.
  • Particle shape: Spherical.
  • Endfitting type: Parker-style.
  • Inner diameter: 6 mm.
  • Length: 150 mm.
  • Packing solvent: THF.
  • System: HPLC.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.