Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters HSPgel Column, 3 μm, 6 mm ID, 150 mm, THF

Catalog No. 50780730 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The HSPgel columns are designed for high-speed GPC analysis. They offer fast, accurate, and precise molecular weight polymer determinations, increase sample throughput, and significantly reduce solvent consumption and disposal costs by allowing analysis with lower volume columns. This leads to total analysis times of 7 minutes or less, increasing sample throughput up to 6x and decreasing solvent usage from 35 to 50 mL to 5 mL or less per run.

  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • Molecular weight range: 1,000 to 4,000,000
  • Particle shape: Spherical
  • Particle size: 3 μm
  • Endfitting type: Parker-style
  • Format: Column
  • Packing solvent: THF
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 6 mm
  • Length: 150 mm
  • Units per package: 1 pk

Catalog No. 50-780-730 Supplier Waters Corp Supplier No. 186001759
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The HSPgel columns are designed for high-speed GPC analysis. They offer fast, accurate, and precise molecular weight polymer determinations, increase sample throughput, and significantly reduce solvent consumption and disposal costs by allowing analysis with lower volume columns. This leads to total analysis times of 7 minutes or less, increasing sample throughput up to 6x and decreasing solvent usage from 35 to 50 mL to 5 mL or less per run.

  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • Molecular weight range: 1,000 to 4,000,000
  • Particle shape: Spherical
  • Particle size: 3 μm
  • Endfitting type: Parker-style
  • Format: Column
  • Packing solvent: THF
  • System: HPLC
  • Technique: GPC
  • Inner diameter: 6 mm
  • Length: 150 mm
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.