Learn More
Aviva Systems Biology Corp HSPH1 Antibody - middle region (ARP52217_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52217_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSPH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HSPH1. Alternate names: DKFZp686M05240, HSP105, HSP105A, HSP105B, KIAA0201, NY-CO-25ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60703