Learn More
Waters Corp Waters XSelect HSS C18 Method Transfer Kit, 100Å, 1.8 to 5 μm
Catalog No. 50791272 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Method transfer kits are designed to preserve the integrity of a separation as it is transferred between UPLC and HPLC platforms. Based on the concept of maintaining column length [L] to particle size [dp] ratio [L/dp], these kits provide a UPLC column with an HPLC column of equivalent selectivity and resolving power. Using the columns calculator, methods can be fully transferred from HPLC to UPLC or from UPLC to HPLC.
  • Designed to preserve separation integrity during transfer between UPLC and HPLC platforms.
  • Maintains column length to particle size ratio [L/dp].
  • Includes a UPLC column with an HPLC column of equivalent selectivity and resolving power.
  • Allows full method transfer from HPLC to UPLC or UPLC to HPLC using the columns calculator.
Catalog No. 50-791-272 Supplier Waters Corp Supplier No. 186004968
Method transfer kits are designed to preserve the integrity of a separation as it is transferred between UPLC and HPLC platforms. Based on the concept of maintaining column length [L] to particle size [dp] ratio [L/dp], these kits provide a UPLC column with an HPLC column of equivalent selectivity and resolving power. Using the columns calculator, methods can be fully transferred from HPLC to UPLC or from UPLC to HPLC.
  • Designed to preserve separation integrity during transfer between UPLC and HPLC platforms.
  • Maintains column length to particle size ratio [L/dp].
  • Includes a UPLC column with an HPLC column of equivalent selectivity and resolving power.
  • Allows full method transfer from HPLC to UPLC or UPLC to HPLC using the columns calculator.