Learn More
Waters Corp Waters Corporation Column, XSelect HSS C18, 100Å, 3.5 μm, 4.6 mm X 50 mm
Supplier:  Waters Corp 186004772
Encompass_Preferred
The HSS C18 is a general-purpose C18 bonded phase column that provides superior peak shape for bases, increased retention (vs. XBridge C18 columns), and exceptional column lifetimes at low pH. The selectivity and retention observed with HSS C18 columns will resemble that of most modern, fully endcapped, high-coverage silica-based C18 columns.
  • Provides superior peak shape for bases.
  • Offers increased retention.
  • Delivers exceptional column lifetimes at low pH.
  • Selectivity and retention resemble modern, fully endcapped, high-coverage silica-based C18 columns.
Catalog No. 50-466-089