Learn More
Waters Corp Waters Corporation Column, C18, 3.5 μm, 4.6 mm X 250 mm, 100 Å
Catalog No. 50466087 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This general-purpose C18 bonded phase column separates complex mixtures of basic and non-basic chemicals at low pH. Using High Strength Silica (HSS) technology, it delivers superior peak shape, increased retention, and exceptional column lifetimes. Adaptable and scalable for UPLC separations, it offers improved polar molecule retention and unique selectivity.
  • Enhanced peak shape for basic chemicals
  • Greater retention
  • Outstanding column lifetimes at low pH
  • Reduced column bleed
  • Outstanding batch-to-batch reproducibility
  • Ultra-efficiencies
  • Seamless column format transferability
  • Improved resolution for complex samples by combining efficiency and selectivity
  • Improved selectivity for acidic, alkaline, and neutral compounds
  • Enhanced retention for basic polar compounds when using low-pH mobile phases
Catalog No. 50-466-087 Supplier Waters Corp Supplier No. 186004770
This general-purpose C18 bonded phase column separates complex mixtures of basic and non-basic chemicals at low pH. Using High Strength Silica (HSS) technology, it delivers superior peak shape, increased retention, and exceptional column lifetimes. Adaptable and scalable for UPLC separations, it offers improved polar molecule retention and unique selectivity.
  • Enhanced peak shape for basic chemicals
  • Greater retention
  • Outstanding column lifetimes at low pH
  • Reduced column bleed
  • Outstanding batch-to-batch reproducibility
  • Ultra-efficiencies
  • Seamless column format transferability
  • Improved resolution for complex samples by combining efficiency and selectivity
  • Improved selectivity for acidic, alkaline, and neutral compounds
  • Enhanced retention for basic polar compounds when using low-pH mobile phases