Learn More
Waters Corp Waters Column, C18, 100 Å, 3.5 μm, 4.6 mm × 150 mm
Supplier:  Waters Corp 186004768
Encompass_Preferred
The HSS C18 is a general-purpose C18 bonded phase column offering superior peak shape for bases, increased retention, and exceptional column lifetimes at low pH. Its selectivity and retention are comparable to modern, fully endcapped, high-coverage silica-based C18 columns.
  • Superior peak shape for basic compounds.
  • Low column bleed and excellent batch-to-batch reproducibility.
  • Ultra-efficiencies and seamless transferability between column formats.
  • Ideal for low pH separations with complex mixtures.
  • Utilizes high strength silica (HSS) technology for increased polar compound retention and mechanical strength under UPLC pressures.
  • Offers alternative analyte selectivity for complex samples, including acidic, alkaline, and neutral compounds.
  • Enhanced retention for basic polar compounds using low-pH mobile phases.
  • Operating pH range: 1 to 8.
Catalog No. 50-466-085