Learn More
Waters Corp Waters Column, C18, 5 μm, 4.6 mm X 150 mm, 100 Å
Supplier:  Waters Corp 186004773
Encompass_Preferred
This C18 bonded phase column provides superior peak shape for bases, increased retention, and exceptional column lifetimes at low pH. Its selectivity and retention resemble modern, fully endcapped, high-coverage silica-based C18 columns, suitable for low pH separations with complex mixtures.
  • Superior peak shape for bases and increased retention
  • Exceptional column lifetimes at low pH
  • Low column bleed and excellent reproducibility
  • Seamless transferability between column formats
  • HSS technology for increased polar compound retention and different selectivity
  • HSS particle provides mechanical strength for UPLC pressures
  • Enhanced resolution for complex samples
  • Enhanced selectivity for acidic, alkaline, and neutral compounds; enhanced retention for basic polar compounds using low-pH mobile phases
Catalog No. 50-466-090